missing translation for 'onlineSavingsMsg'
Learn More

Nimesulide

Highly selective cyclooxygenase-2 inhibitor which induces apoptosis | CAS: 51803-78-2 | C13H12N2O5S | 308.308 g/mol

Supplier:  Thermo Scientific Chemicals J6369906

 View more versions of this product

Catalog No. AAJ6369906

Only null left
Explore available promotions

Description

Description

Nimesulide are used for the treatment of acute pain, the symptomatic treatment of osteoarthritis and primary dysmenorrhoea, . Nimesulide was as effective as paracetamol in reducing fever, local pain, and general discomfort. It was effective in reducing pain associated with osteoarthritis, cancer, thrombophlebitis, oral surgery in adults, otorhinolaryngological diseases and traumatic injury. Nimesulide is demonstrated to suppress azoxymethane-induced carcinogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (<50 µg/ml), 1:10 DMSO:PBS (pH 7.2) (<200 µg/ml), ethanol (1 mg/ml), DMSO (15 mg/ml), DMF (15 mg/ml), chloroform, dichloromethane, acetone (freely soluble), and 1N NaOH.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

Nimesulide
145°C to 147°C
C13H12N2O5S
5 g
14,6548
Soluble in water (<50 μg/ml),1:10 DMSO:PBS (pH 7.2) (<200 μg/ml),ethanol (1mg/ml),DMSO (15mg/ml),DMF (15mg/ml),chloroform,dichloromethane,acetone (freely soluble),and 1N NaOH.
CS(=O)(=O)NC1=C(C=C(C=C1)[N+](=O)[O-])OC2=CC=CC=C2
308.308
CHEBI:44445
Needle-like Crystals
51803-78-2
Yellow
MFCD00079470
UN2811
nimesulide, mesulid, n-4-nitro-2-phenoxyphenyl methanesulfonamide, flogovital, sulidene, nimed, 4-nitro-2-phenoxymethanesulfonanilide, nisulid, nimesulidum inn-latin, nimesulida inn-spanish
HYWYRSMBCFDLJT-UHFFFAOYSA-N
N-(4-nitro-2-phenoxyphenyl)methanesulfonamide
4495
308.31

Chemical Identifiers

51803-78-2
308.308
HYWYRSMBCFDLJT-UHFFFAOYSA-N
4495
N-(4-nitro-2-phenoxyphenyl)methanesulfonamide
C13H12N2O5S
MFCD00079470
nimesulide, mesulid, n-4-nitro-2-phenoxyphenyl methanesulfonamide, flogovital, sulidene, nimed, 4-nitro-2-phenoxymethanesulfonanilide, nisulid, nimesulidum inn-latin, nimesulida inn-spanish
CHEBI:44445
CS(=O)(=O)NC1=C(C=C(C=C1)[N+](=O)[O-])OC2=CC=CC=C2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 257-431-4

RTECSNumber : PB0970000

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only