missing translation for 'onlineSavingsMsg'
Learn More

Nimesulide

Catalog No. p-7042769
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6369906 5 g
AAJ6369914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6369906 Supplier Thermo Scientific Chemicals Supplier No. J6369906

Highly selective cyclooxygenase-2 inhibitor which induces apoptosis | CAS: 51803-78-2 | C13H12N2O5S | 308.308 g/mol

Nimesulide are used for the treatment of acute pain, the symptomatic treatment of osteoarthritis and primary dysmenorrhoea, . Nimesulide was as effective as paracetamol in reducing fever, local pain, and general discomfort. It was effective in reducing pain associated with osteoarthritis, cancer, thrombophlebitis, oral surgery in adults, otorhinolaryngological diseases and traumatic injury. Nimesulide is demonstrated to suppress azoxymethane-induced carcinogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (<50 µg/ml), 1:10 DMSO:PBS (pH 7.2) (<200 µg/ml), ethanol (1 mg/ml), DMSO (15 mg/ml), DMF (15 mg/ml), chloroform, dichloromethane, acetone (freely soluble), and 1N NaOH.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 51803-78-2
Molecular Formula C13H12N2O5S
Molecular Weight (g/mol) 308.308
MDL Number MFCD00079470
InChI Key HYWYRSMBCFDLJT-UHFFFAOYSA-N
Synonym nimesulide, mesulid, n-4-nitro-2-phenoxyphenyl methanesulfonamide, flogovital, sulidene, nimed, 4-nitro-2-phenoxymethanesulfonanilide, nisulid, nimesulidum inn-latin, nimesulida inn-spanish
PubChem CID 4495
ChEBI CHEBI:44445
IUPAC Name N-(4-nitro-2-phenoxyphenyl)methanesulfonamide
SMILES CS(=O)(=O)NC1=C(C=C(C=C1)[N+](=O)[O-])OC2=CC=CC=C2

Specifications

Melting Point 145°C to 147°C
Color Yellow
Quantity 5 g
UN Number UN2811
Merck Index 14,6548
Solubility Information Soluble in water (<50 μg/ml),1:10 DMSO:PBS (pH 7.2) (<200 μg/ml),ethanol (1mg/ml),DMSO (15mg/ml),DMF (15mg/ml),chloroform,dichloromethane,acetone (freely soluble),and 1N NaOH.
Formula Weight 308.31
Physical Form Needle-like Crystals
Chemical Name or Material Nimesulide
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 257-431-4
RTECSNumber PB0970000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only