accessibility menu, dialog, popup

UserName

CAS RN 51803-78-2

IUPAC Name: N-(4-nitro-2-phenoxyphenyl)methanesulfonamide
Synonyms: nimesulide
Molecular Weight (g/mol): 308.31
Molecular Formula: C13H12N2O5S
InChi Key: HYWYRSMBCFDLJT-UHFFFAOYSA-N
SMILES: CS(=O)(=O)NC1=CC=C(C=C1OC1=CC=CC=C1)[N+]([O-])=O
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)