missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nile Red, 99%, pure

Numéro de catalogue. p-172452
Change view
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC415711000 100 mg Glass bottle
AC415710010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC415711000 Fournisseur Thermo Scientific Chemicals Code fournisseur 415711000
Il en reste null

CAS: 7385-67-3 | C20H18N2O2 | 318.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 7385-67-3
Molecular Formula C20H18N2O2
Molecular Weight (g/mol) 318.38
MDL Number MFCD00011639
InChI Key VOFUROIFQGPCGE-UHFFFAOYSA-N
Synonym Nile blue A oxazone
PubChem CID 65182
ChEBI CHEBI:52169
SMILES CCN(CC)C1=CC=C2N=C3C(OC2=C1)=CC(=O)C1=CC=CC=C31

Spécifications

Melting Point 203.0°C to 205.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 27, II, 468
Merck Index 15, 6628
Solubility Information Solubility in water: almost insoluble. Other solubilities: soluble in acetone, acetonitrile, chloroform, thf,, ethanol, dmso, cs2, benzene, nitrobenzene, pyridin, e, hexane, diethyl ether
Infrared Spectrum Authentic
IUPAC Name 9-(diethylamino)benzo[a]phenoxazin-5-one
Formula Weight 318.38
Percent Purity 99%
Grade Pure
Packaging Glass bottle
UV_Vis 98.5% min.
UV Spectrometry Complies
Color Maroon to Red
Quantity 100 mg
Physical Form Powder
Chemical Name or Material Nile Red, 99%
Afficher plus de résultats Afficher moins de résultats
EINECSNumber 230-966-0
TSCA TSCA

RUO – Research Use Only