missing translation for 'onlineSavingsMsg'
Learn More

Nigrosine (Water soluble), TCI America™

Catalog No. N078225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N078225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N078225G Supplier TCI America Supplier No. N078225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 8005-03-6
Molecular Formula C22H14N6Na2O9S2
Molecular Weight (g/mol) 616.49
InChI Key HKBVRFLHNUEVRO-HMDKGIQMSA-L
Synonym amido black 10b, acid black 1, buffalo black nbr, naphthol blue black, aniline blue black, tech, nigrosine water soluble, 4-amino-5-hydroxy-3-4-nitrophenyl azo-6-phenylazo naphthalene-2,7-disulphonic acid, sodium salt, 2,7-naphthalenedisulfonic acid, 4-amino-5-hydroxy-3-4-nitrophenyl azo-6-phenylazo-, sodium salt, 3-4-nitrophenylazo-4-amino-5-oxo-6-2-phenylhydrazono-5,6-dihydro-2,7-naphthalenedisulfonic acid disodium salt, disodium 3e-5-amino-6-4-nitrophenyl diazenyl-4-oxo-3-phenylhydrazinylidene naphthalene-2,7-disulfonate
PubChem CID 44134531
IUPAC Name disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color Black
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Nigrosine (Water soluble)
EINECSNumber (5)-1876
RTECSNumber GC4762250
TSCA Yes