missing translation for 'onlineSavingsMsg'
Learn More

Niflumic acid, 99+%

Catalog No. AAJ6048918
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6048918 50 g
AAJ6048906 5 g
AAJ6048909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6048918 Supplier Thermo Scientific Chemicals Supplier No. J6048918

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4394-00-7 | C13H9F3N2O2 | 282.22 g/mol

It is used as a selective Cox-2 inhibitor and GPR35 activator. Also activates human TRPA1 inducibly expressed in HEK293 cells; displays agonist activity at GPR35.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a selective Cox-2 inhibitor and GPR35 activator. Also activates human TRPA1 inducibly expressed in HEK293 cells; displays agonist activity at GPR35.

Solubility
Soluble in ethanol (∼50 mg/ml), acetone (50 mg/ml, Clear to Slightly Hazy, Yellow), methanol (∼50 mg/ml), DMSO (56 mg/ml at 25°C), and acetonitrile (∼50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Hygroscopic. Keep away fom strong oxidizing agents.

Chemical Identifiers

CAS 4394-00-7
Molecular Formula C13H9F3N2O2
Molecular Weight (g/mol) 282.22
MDL Number MFCD00010569
InChI Key JZFPYUNJRRFVQU-UHFFFAOYSA-N
Synonym niflumic acid, nifluril, landruma, forenol, actol, acido niflumico, nifluminic acid, acide niflumique, acidum niflumicum, acide niflumique inn-french
PubChem CID 4488
ChEBI CHEBI:34888
IUPAC Name 2-{[3-(trifluoromethyl)phenyl]amino}pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=CN=C1NC1=CC=CC(=C1)C(F)(F)F

Specifications

Melting Point 203°C to 204°C
Color Yellow
Quantity 50 g
UN Number UN2811
Sensitivity Light sensitive
Merck Index 14,6531
Solubility Information Soluble in ethanol (∽50mg/ml),acetone (50mg/ml,Clear to Slightly Hazy,Yellow),methanol (∽50mg/ml),DMSO (56mg/ml at 25°C),and acetonitrile (∽50mg/ml).
Formula Weight 282.22
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material Niflumic acid
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H312-H332-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 224-516-2
RTECSNumber QT2999100
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only