missing translation for 'onlineSavingsMsg'
Learn More

Nifedipine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012180
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012180 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012180 Supplier Merck Emd Millipore Supplier No. PHR12901G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Nifedipine belongs to the class of calcium channel antagonist drugs and finds a wide use as an antihypertensive and an antianginal agent. It can also be used as a coronary vasodilator.

Specifications

CAS 21829-25-4
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C17H18N2O6
MDL Number MFCD00057326
Synonym Nifedipine; 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic acid dimethyl ester
Type Certified Reference Material
Quantity 1 g
Formula Weight 346.33
Grade Certified Reference Material