missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic acid, MilliporeSigma™ Supelco™

Catalog No. 111014808
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014808 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014808 Supplier Merck Emd Millipore Supplier No. 47864

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nicotinic acid is a water soluble, vitamin belonging to B complex group of vitamins, which can be used as a broad spectrum lipid lowering drug. Its mode of action basically involves the inhibition of the hormone-sensitive triglyceride lipase and thereby, decreasing the lipolysis in adipose tissue.

Pharmaceutical (small molecule) Vitamins, Nutraceuticals and Natural Products

Specifications

Packaging Ampule
CAS 59-67-6
Flash Point 193°C
Physical Form Neat
Linear Formula C6H5 NO2
MDL Number MFCD00006391
Melting Point 236°C to 239°C
Synonym 3-Picolinic acid; Niacin; Pellagra preventive factor; Pyridine-3-carboxylic acid; Vitamin B3
Type Analytical Standard
Quantity 1000 mg
Formula Weight 123.11
Grade Analytical Standard
Show More Show Less