missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Nicotinamide

Catalog No. 410650
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
410650 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 410650 Supplier Tocris Bioscience™ Supplier No. 4106/50

PARP-1 inhibitor

Nicotinamide is an anti-inflammatory agent. Inhibitor of poly(ADP-ribose) polymerase (PARP-1) enzymes. NAD+ precursor. Promotes differentiation of mesenchymal stem cells to insulin producing cells when used in combination with growth factors and high glucose concentration. Nicotinamide also acts as a SIRT1 inhibitor and promotes the expansion of hematopoietic progenitor cells

For more information about how Nicotinamide may be used, see our protocol:

3D Culture of Lung Alveolar Cells

Chemical Identifiers

CAS 98-92-0
Molecular Formula C6H6N2O

Specifications

Quantity 50 mg
Formula Weight 122.12
Purity >99%
Molecular Weight (g/mol) 122.127
Chemical Name or Material Nicotinamide
Recommended Storage Store at Room Temperature