missing translation for 'onlineSavingsMsg'
Learn More

Nicotinamide Impurity E, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023001
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023001 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023001 Supplier Merck Emd Millipore Supplier No. PHR2090100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 1986-81-8
Melting Point 291°C to 293°C (decomposition) (literature)
Flash Point Not Applicable
Molecular Formula C6H6N2O2
Linear Formula C6H6N2O2
Quantity 100 mg
MDL Number MFCD00006202
Synonym Nicotinamide N-oxide
Formula Weight 138.12
Grade Certified Reference Material