missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinamide Impurity D Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 1453-82-3 |
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C6H6N2 O |
| MDL Number | MFCD00006432 |
| Melting Point | 155°C to 157°C |
| Synonym | Isonicotinamide; Pyridine-4-carboxylic acid amide |
| Type | Certified Reference Material |
| Quantity | 100 mg |
| Formula Weight | 122.12 |
| Grade | Certified Reference Material |