missing translation for 'onlineSavingsMsg'
Learn More

Nickel wire, 0.15mm (0.006in) dia, annealed, Nickel 200, approximately 99% (metals basis)

Catalog No. AA41785I1
Change view
Click to view available options
Quantity:
250 m
1000 m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41785I1 1000 m
AA41785H7 250 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA41785I1 Supplier Thermo Scientific Chemicals Supplier No. 041785I1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Form: Approx.0.16g/m Temper: soft | CAS: 7440-02-0 | Ni | 58.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Color Silver to Gray
Appearance Silver gray wire
Quantity 1000 m
Odor Odorless
Form Wire
Assay Percent Range approx.99%, Nickel 200 (Metals basis)
Chemical Name or Material Nickel
Hazard Category H317-H351-H372
Precautionary Statement P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 231-111-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only