missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) p-Toluenesulfonate Hexahydrate 98.0+%, TCI America™

Numéro de catalogue. T02765G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T02765G 5 g
T027625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. T02765G Fournisseur TCI America Code fournisseur T02765G
Il en reste null

Identifiants chimiques

CAS 6944-05-4
Molecular Formula C7H8NiO3S
Molecular Weight (g/mol) 230.89
MDL Number MFCD00070542
InChI Key KXIBSBLPGLHJNY-UHFFFAOYSA-N
Synonym p-Toluenesulfonic Acid Nickel(II) Salt
PubChem CID 54603762
IUPAC Name 4-methylbenzenesulfonic acid;nickel
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.[Ni]

Spécifications

Color Green-Yellow
Quantity 5 g
Formula Weight 401.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nickel(II) p-Toluenesulfonate Hexahydrate