Learn More
Nickel(II) nitrate hexahydrate, Puratronic™, 99.9985% (metals basis)
Description
It is used as a precursor in the manufacturing of other nickel salts and supported nickel catalysts. It finds use as a catalyst in organic synthesis. Further it is used in electroplating and as a mordent in textile dyeing. It is a precursor for preparing nickel sulfide-reduced graphene oxide (RGO) composite powders with high initial capacities and good cycling performance. It can be employed in the preparation of nickel and nickel oxide nanocrystals impregnated O-MWCNTs (oxygen-functionalized multi-walled carbon nanotubes) with potential applications in semiconductor industries. It finds use in improving cycling stability ad rate capability of Li2MnO3.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 137°C |
| Melting Point | 56.7°C |
| Color | Green |
| Physical Form | Crystalline Aggregates |
| Quantity | 250 g |
| Linear Formula | Ni(NO3)2·6H2O |
| UN Number | UN2725 |
| Merck Index | 14,6513 |
| Solubility Information | Very soluble in water. Soluble in ethanol. |
| IUPAC Name | nickel(2+) hexahydrate dinitrate |
| Show More |
Safety and Handling
| Hazard Category | H272-H302+H332-H315-H317-H318-H334-H335-H341-H350i-H360D-H372 |
| Hazard Statement | GHS H Statement H272-H334-H350-H360-H372-H341-H318-H302-H332-H315-H317 May intensify fire; oxidizer. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Causes serious eye damage. Harmful if swallowed. Harmful if inhaled. Causes skin irritation. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P210-P220-P221-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P342+P311-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NICKEL NITRATE |
| EINECSNumber | 236-068-5 |
| RTECSNumber | QR7300000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only