missing translation for 'onlineSavingsMsg'
Learn More

Nickel (II) Chloride Hexahydrate, Purified, Reagents

Catalog No. 01337158
Change view
Click to view available options
Quantity: 500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01337158 Quantity: 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01337158 Supplier Labchem Inc Supplier No. C2255500500A5

Nickel (II) Chloride Hexahydrate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7791-20-0
Molecular Formula Cl2H12NiO6
Molecular Weight (g/mol) 237.68
InChI Key LAIZPRYFQUWUBN-UHFFFAOYSA-L
Synonym Nickel(II) Chloride Hexahydrate, Nickelous Chloride Hexahydrate
IUPAC Name nickel(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ni++]

Specifications

Boiling Point 973°C
Color Green
Physical Form Solid
Quantity 500 g
Grade Pure
Packaging Poly Bottle
Specific Gravity 3.55
Chemical Name or Material Nickel (II) Chloride Hexahydrate, Purified