missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) chloride hexahydrate, Puratronic™, 99.999% (metals basis)
Description
Used in the manufacture of all kinds of nickel compounds, preservatives and chemical reaction catalyst. Also used in electroplating, analytical chemistry, used for analysis reagent and in gas masks to absorb NH3. It has also been utilized in environmental studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 140°C-H2O |
| Color | Green |
| Physical Form | Crystalline |
| Quantity | 100 g |
| Odor | Odorless |
| Assay Percent Range | 99.999% (Metals Basis) |
| Density | 1.92 |
| Solubility Information | Soluble in water (2540g/L) and ethanol. |
| Formula Weight | 237.71 (129.60 Anhydrous) |
| Chemical Name or Material | Nickel(II) chloride hexahydrate |
Safety and Handling
| Hazard Category | H301+H331-H315-H317-H319-H334-H341-H350i-H360D-H372 |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P330-P333+P313-P362-P501c |
| DOTInformation | DOT Class: 6.1, Packaging Group: III |
| EINECSNumber | 231-743-0 |
| RTECSNumber | QR6480000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only