missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) chloride hexahydrate, 98+%

Catalog No. AC270512500
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC270512500 250 g Glass bottle
AC270510010 1 kg Glass bottle
AC270510050 5 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC270512500 Supplier Thermo Scientific Chemicals Supplier No. 270512500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7791-20-0 | Cl2Ni·6H2O | 237.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7791-20-0
Molecular Formula Cl2Ni·6H2O
Molecular Weight (g/mol) 237.71

Specifications

Color Green
pH 4 to 6 (5% soln. at 20°C)
Physical Form Crystalline Powder and/or Crystals
Quantity 250 g
Packaging Glass bottle
Assay Percent Range 98% min. (Complexometry)
Linear Formula NiCl2·6H2O
Fieser 14,351; 15,96; 17,86
Merck Index 15,659
Solubility Information Solubility in water: 2540g/L water (20°C). Other solubilities: soluble in ammonium hydroxide, soluble in alcohol
Formula Weight 237.71
Percent Purity ≥98%
Chemical Name or Material Nickel(II) chloride hexahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.<br
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid relea

RUO – Research Use Only