missing translation for 'onlineSavingsMsg'
Learn More

Nickel Chloride Hexahydrate, (Crystalline),MP Biomedicals

Catalog No. ICN15582590
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15582590 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15582590 Supplier MP Biomedicals Inc Supplier No. 0215582590

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cell Culture Reagent

Chemical Identifiers

CAS 7791-20-0
Molecular Formula NiCl2 · 6H2O

Specifications

Physical Form Crystalline
Quantity 500 g
Formula Weight 237.7
Chemical Name or Material Nickel Chloride Hexahydrate
Recommended Storage Room Temperature