missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Chloride, Hexahydrate, Crystal, Purified, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystalline Powder or Needles |
| Quantity | 45 kg |
| Molecular Weight (g/mol) | 237.71 |
| Grade | Purified |
| Packaging | Amber Glass Bottle |
| Chemical Name or Material | Nickel Chloride, Hexahydrate, Crystal |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.