missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, AAS standard solution, Specpure™ Ni 1000μg/mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Liquid |
| Concentration | Ni in 5% HNO3 |
| UN Number | UN3264 |
| Color | Colorless |
| Quantity | 100 mL |
| Grade | Specpure |
| Chemical Name or Material | Nickel, AAS standard solution, Ni 1000μg/mL |
Safety and Handling
| Hazard Category | H290-H314-H335-H350i-H360D |
| Hazard Statement | GHS H Statement H314-H317-H208 Causes severe skin burns and eye damage. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P234-P260-P264b-P271-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only