missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel AA Standard, certifié, 1000 ppm ±10 ppm (1 mL = 1 mg Ni), LabChem™
Spécifications
Spécifications
| Nom ou substance chimique | Nickel AA Standard |
| Nom Remarque | 1000 ppm (1 ml = 1 mg Ni) |
| Emballage | Bouteille en polyéthylène |
| CAS % maximal | 95.76, 3.74, 0.5 |
| Désignation | Test de réussite |
| Viscosité | 0.99cSt |
| Densité | 1.02g/mL |
| Forme physique | Liquide |
| Concentration | 1000 ppm ±10 ppm (1 ml = 1 mg de Ni) |
| Formule linéaire | Ni(NO3)2·6H2O |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement May be corrosive to metals. Causes severe skin burns and eye damage. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause cancer. May damage fertility or the unborn child. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Do not handle until all safety precautions have been read and understood. Keep only in original container. Do not breathe mist. Wear protective gloves, protective clothing, eye protection, face protection. Wear respiratory protection. Wash exposed skin thoroughly after handling. Contaminated work clothing should not be allowed out of the workplace. Absorb spillage to prevent material damage. If swallowed: Rinse mouth. Do not induce vomiting. Immediately call a poison center/doctor. If on skin (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a poison center/doctor. Wash contaminated clothing before reuse. If inhaled: Remove person to fresh air and keep comfortable for breathing. Immediately call a poison center/doctor. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Store in corrosive resistant container with a resistant inner liner. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Température ambiante |