missing translation for 'onlineSavingsMsg'
Learn More

[NH2Me2][(RuCl((S)-xylbinap))2(mu-Cl)3], TCI America™

Catalog No. N1020200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1020200MG 200 mg
N10201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N1020200MG Supplier TCI America Supplier No. N1020200MG

Chemical Identifiers

CAS 944451-10-9
Molecular Formula C106H104Cl5NP4Ru2
Molecular Weight (g/mol) 1895.29
MDL Number MFCD09753025
InChI Key KEZRTMOWSKGUPV-UHFFFAOYSA-J
Synonym Dimethylammonium Dichlorotri(mu-chloro)bis[(S)-(-)-2,2′C-bis[di-(3,5-xylyl)phosphino]-1,1′C-binaphthyl]diruthenate(II)
PubChem CID 131675154
IUPAC Name [1-[2-bis(3,5-dimethylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(3,5-dimethylphenyl)phosphane;N-methylmethanamine;ruthenium(2+);tetrachloride;hydrochloride
SMILES CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.CNC.Cl.[Cl-].[Cl-].[C

Specifications

Quantity 200 mg
Formula Weight 1895.29
Physical Form Crystalline Powder
Chemical Name or Material [NH2Me2][(RuCl((S)-xylbinap))2(mu-Cl)3]