missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nextteq™ Gastec™ Detector Tubes: Phenol
Description
Gastec tubes offer accurate, effective readings of more than 500 gases and vapors. Each glass detector tube contains a reagent which reacts with a specific gas or vapor, yielding a stain that is measured against calibrations printed on the tube. The unique absorbing media produce extremely clear lines of stain demarcation for precise reading, and are stable, with a shelf-life of up to two years, depending on the application. (See below for tube selection of specific gases and vapors.) Sold in packs of 10.
Specifications
Specifications
| Components Measured | Phenol |
| Type | Gas Detection Tube |
| Measurement Range | 0.4 to 187ppm |
| No. of Strokes | 0.5 to 4 |
| Quantity | 10 |
| Response Time | 45 sec. to 6 min. |
| Temperature Range (English) | 32°F to 104°F |
| Temperature Range (Metric) | 0°C to 40°C |