missing translation for 'onlineSavingsMsg'
Learn More

Nextteq™ Gastec™ Detector Tubes: Carbon Monoxide

Catalog No. 19026242
Change view
Click to view available options
Measurement Range:
5 to 50
Response Time:
4 min.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Range Response Time
19026242 5 to 50 4 min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19026242 Supplier Nextteq™ Supplier No. 1LL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Each tube contains a reagent that reacts with a specific gas/vapor, yielding a stain that is measured against calibrations printed on the tube.

Gastec tubes offer accurate, effective readings of more than 500 gases and vapors. Each glass detector tube contains a reagent which reacts with a specific gas or vapor, yielding a stain that is measured against calibrations printed on the tube. The unique absorbing media produce extremely clear lines of stain demarcation for precise reading, and are stable, with a shelf-life of up to two years, depending on the application. (See below for tube selection of specific gases and vapors.) Sold in packs of 10.

Specifications

Components Measured Carbon Monoxide
For Use With (Equipment) GV-100/GV-110 Ga Sampling Pump
Type Gas Detection Tube
Measurement Range 5 to 50
No. of Strokes 2
Quantity 10
Response Time 4 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C