Learn More
New England Biolab Ltd NEBNext Multi Small RNA Lib Prep IL Set2 24 reactions
Numéro de catalogue. 50591163
Encompass
The novel NEBNext Small RNA workflow has been optimized to minimize adaptor-dimers while producing high-yield, high-diversity libraries. The NEBNext Small RNA Library Prep Set for Illumina (Set 1) includes adaptors and multiplex primers with 12 indices, to enable multiplexing.
Numéro de catalogue. 50591163 Fournisseur New England Biolab Ltd Code fournisseur E7580S
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
The novel NEBNext Small RNA workflow has been optimized to minimize adaptor-dimers while producing high-yield, high-diversity libraries. The NEBNext Small RNA Library Prep Set for Illumina (Set 1) includes adaptors and multiplex primers with 12 indices, to enable multiplexing.