NEBNext Multiplex Oligos provide adaptors and primers to enable high yield multiplex Illumina library production. The unique hairpin loop structure of the NEBNext Adaptor minimizes adaptor-dimer formation, and NEBNext index PCR primers enable index incorporation during library amplification. This kit includes 8 i5 index primers and 12 i7 index primers for dual indexing. Additional NEBNext Multiplex Oligos products are available for different indexing strategies.