missing translation for 'onlineSavingsMsg'
Learn More

New Methylene Blue, Brecher Formulation, for Staining Reticulocytes in Peripheral Blood, Ricca Chemical

$124.83 - $314.17

Chemical Identifiers

CAS 6586-05-6
Molecular Formula C18H22Cl3N3SZn
Molecular Weight (g/mol) Mixture
InChI Key GGKWUSWRGRCLEV-UHFFFAOYSA-K
PubChem CID 3083969
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5280-4
View Documents
Ricca Chemical Company
52804
120 mL
Each for $124.83
Only null left
 
5280-16
View Documents
Ricca Chemical Company
528016
16 oz.
Each for $314.17
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6586-05-6
Mixture
3083969
C18H22Cl3N3SZn
GGKWUSWRGRCLEV-UHFFFAOYSA-K

Specifications

6586-05-6 , 6487-48-5 , 1934-16-3
100.0,1.6,0.39
GGKWUSWRGRCLEV-UHFFFAOYSA-K
3083969
Natural Poly Bottle
120 mL
96.0,1.54,0.37
C18H22Cl3N3SZn
Mixture
Laboratory
Dark Blue
New Methylene Blue, Brecher
Videos
SDS
Documents

Documents