missing translation for 'onlineSavingsMsg'
Learn More
Learn More
New Methylene Blue, Brecher Formulation, for Staining Reticulocytes in Peripheral Blood, Ricca Chemical
Specifications
Specifications
| CAS Min % | 96.0, 1.54, 0.37 |
| CAS Max % | 100.0, 1.6, 0.39 |
| Packaging | Natural Poly Bottle |
| Color | Dark Blue |
| Quantity | 120 mL |
| Grade | Laboratory |
| Chemical Name or Material | New Methylene Blue, Brecher |