missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Neutral Red

Catalog No. AAJ6305414
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6305414 25 g
AAJ6305403 1 g
AAJ6305406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6305414 Supplier Thermo Scientific Chemicals Supplier No. J6305414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nuclear counterstain useful in pH range 6.8 to 8.0 | CAS: 553-24-2 | C15H17ClN4 | 288.779 g/mol

Nuclear counterstain useful in pH range 6.8 - 8.0

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 553-24-2
Molecular Formula C15H17ClN4
Molecular Weight (g/mol) 288.779
MDL Number MFCD00012651
InChI Key PGSADBUBUOPOJS-UHFFFAOYSA-N
Synonym Basic Red 5; C.I. 50040
PubChem CID 11105
ChEBI CHEBI:86370
IUPAC Name 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride
SMILES CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl

Specifications

Melting Point 290°C (decomposition)
Beilstein 3918740
Merck Index 14,6488
Color Black to Green
Quantity 25 g
Physical Form Powder
Formula Weight 288.78
Chemical Name or Material Neutral Red
EINECSNumber 209-035-8
RTECSNumber SG1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only