missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurocan, Rabbit anti-Rat, Polyclonal, Millipore Sigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABT138225UL
Description
This rabbit polyclonal Anti-Neurocan, Cat. No. ABT1382, is a rabbit polyclonal antibody that detects Neurocan core protein and has been tested for use in Immunofluorescence.
Neurocan core protein (UniProt: P55067; also known as 245 kDa early postnatal core glycoprotein, Chondroitin sulfate proteoglycan 3) is encoded by the Ncan (also known as Cspg3) gene in rat. Neurocan is a secreted protein of the aggrecan/versican proteoglycan family that is found in early postnatal and adult brain. During early postnatal development neurocan accounts for at least 20% of the protein of the soluble brain proteoglycans. Neurocan mostly contains chondroitin sulfate, but also has N-linked and O-linked oligosaccharides. It is shown to be involved in the modulation of cell adhesion and migration and regulates neurite outgrowth. Neurocan interact with several cell surface molecules, such as N-CAM, L1/Ng-CAM, etc., and modulates the cell-binding and neurite outgrowth promoting activates of these molecules. Neurocan is synthesized with a 22 aa signal peptide that is subsequently cleaved to produce the mature form. Neurocan contains one C-type lectin domain, two EGF-like domains, one Ig-like V type domain, two link domains and one Sushi (CCP/SCR) domain. (Ref.: Rauch, U., et al. (2001) Cell Mol. Life Sci. 58(12-13):1842-56; Zhou, XH et al. (2001). Mol. Cell Biol. 21(17): 5970-78).Spécifications
| Neurocan | |
| Polyclonal | |
| Rabbit polyclonal antiserum with 0.05% sodium azide. | |
| Ncan;Cspg3 | |
| Full-length Neurocan isolated from 7-day embryonic rat brain and purified by monoclonal antibody immunoaffinity chromatography. | |
| Unpurified | |
| RUO | |
| Primary | |
| Rat | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| Neurocan core protein;245 kDa early postnatal core glycoprotein;Chondroitin sulfate proteoglycan 3 | |
| Rabbit | |
| 135.55 kDa calculated. | |
| 25 μL | |
| Cell Structure | |
| This rabbit polyclonal antibody detects Neurocan in fetal and adult rat brain. | |
| Stable for 1 year at -20°C from date of receipt. |