missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Neuro2a Whole Cell Lysate
Description
The Neuro2a Whole Cell Lysate is derived from mouse. It has been validated for the following applications: Western Blot.
Specifications
Specifications
| Quantity | 0.25 mg |
| Storage Buffer | 1X Laemmli Sample Buffer [31.5 mM Tris HCl, 1% SDS, 10% Glycerol, 0.005% bromophenol blue, pH 6.8] |
| Purity | Protein |
| Cellular Fraction | Whole |
| Concentration | 5.0 mg/mL |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| For Use With (Application) | Western Blot |
| Tissue | Brain |
For Research Use Only