missing translation for 'onlineSavingsMsg'
Learn More

Neu5Troc[1Me,4789Ac]alpha(2-3)Gal[26Bn]-beta-MP 98.0+%, TCI America™

Catalog No. M17291G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M17291G Supplier TCI America Supplier No. M17291G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 610763-72-9
Molecular Formula C48H56Cl3NO20
Molecular Weight (g/mol) 1073.313
InChI Key HNINFVMNWIPTPG-IJDKSFGZSA-N
PubChem CID 56973700
IUPAC Name methyl 4-acetyloxy-2-[5-hydroxy-2-(4-methoxyphenoxy)-3-phenylmethoxy-6-(phenylmethoxymethyl)oxan-4-yl]oxy-6-[(1S,2R)-1,2,3-triacetyloxypropyl]-5-(2,2,2-trichloroethoxycarbonylamino)oxane-2-carboxylate
SMILES CC(=O)OCC(C(C1C(C(CC(O1)(C(=O)OC)OC2C(C(OC(C2OCC3=CC=CC=C3)OC4=CC=C(C=C4)OC)COCC5=CC=CC=C5)O)OC(=O)C)NC(=O)OCC(Cl)(Cl)Cl)OC(=O)C)OC(=O)C

Specifications

Color White
Quantity 1 g
Formula Weight 1073.31
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Neu5Troc[1Me,4789Ac]alpha(2-3)Gal[26Bn]-beta-MP
TSCA No
Recommended Storage Refrigerator