missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NERL™ High Purity Water

Catalog No. 23249590
Change view
Click to view available options
Quantity:
1 gal.
5 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23249590 5 gal.
23249589 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
  • For in vitro diagnostic and laboratory use only
  • Not for infusion, injection or irrigation
Catalog No. 23249590 Supplier Thermo Scientific™ Supplier No. J9805
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • For in vitro diagnostic and laboratory use only
  • Not for infusion, injection or irrigation

Improve control in critical diagnostic procedures by using Thermo Scientific™ NERL™ High Purity Water in place of deionized or distilled water.

Improve control in critical diagnostic procedures by using Thermo Scientific™ NERL™ High Purity Water in place of deionized or distilled water. This product meets CAP/CLSI specifications for Clinical Laboratory Reagent Grade Water (CLRW) at time of manufacture.

Typically prepared at 18 megohm/cm specific resistance using reverse osmosis, mixed bed deionization, activated carbon filtration and final filtration to 0.2 micron, NERL high purity water is essentially free from organic and inorganic particulate and soluble contaminants.

Contains no preservatives

Ordering Information:

Certificate of analysis available for each case

Specifications

pH 5 to 7
Grade NERL
Quantity 5 gal.
Recommended Storage 15° to 30°C

General Purpose - For in vitro diagnostic or laboratory use only. Not for infusion, injection or irrigation.