missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Trifluoroacetyl-L-lysine 98.0+%, TCI America™

Catalog No. T28155G
missing translation for 'changeView'
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28155G 5 g
T281525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. T28155G Supplier TCI America missing translation for 'supplierNo' T28155G
Only null left

Chemical Identifiers

CAS 10009-20-8
Molecular Formula C8H13F3N2O3
Molecular Weight (g/mol) 242.20
MDL Number MFCD00037223
InChI Key PZZHRSVBHRVIMI-MLHKIVSYNA-N
PubChem CID 7009573
ChEBI CHEBI:61064
IUPAC Name (2S)-2-amino-6-(2,2,2-trifluoroacetamido)hexanoic acid
SMILES N[C@@H](CCCCNC(=O)C(F)(F)F)C(O)=O

Specifications

Melting Point 254°C
Color White
Quantity 5 g
Synonym h-lys tfa-oh, n-6-trifluoroacetyl-l-lysine, n6-trifluoroacetyl-l-lysine, n6-trifluoroaccety-l-lysine, epsilon-tfa-lysine, nepsilon-trifluoroacetyl-l-lysine, l-lysine, n6-trifluoroacetyl, n 6-trifluoroacetyl-l-lysine, n-, a-trifluoroacetyl-l-lysine
Formula Weight 242.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nepsilon-Trifluoroacetyl-L-lysine
TSCA No
Recommended Storage Refrigerator