missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Trifluoroacetyl-L-lysine, 97%

CAS: 10009-20-8 | C8H13F3N2O3 | 242.20 g/mol

$46.84 - $121.36

Chemical Identifiers

CAS 10009-20-8
Molecular Formula C8H13F3N2O3
Molecular Weight (g/mol) 242.20
MDL Number MFCD00037223
InChI Key PZZHRSVBHRVIMI-MLHKIVSYNA-N
Synonym h-lys tfa-oh, n-6-trifluoroacetyl-l-lysine, n6-trifluoroacetyl-l-lysine, n6-trifluoroaccety-l-lysine, epsilon-tfa-lysine, nepsilon-trifluoroacetyl-l-lysine, l-lysine, n6-trifluoroacetyl, n 6-trifluoroacetyl-l-lysine, n-, a-trifluoroacetyl-l-lysine
PubChem CID 7009573
ChEBI CHEBI:61064
SMILES N[C@@H](CCCCNC(=O)C(F)(F)F)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAH3705106
View Documents Promotion Details
N/A 5 g
Each for $46.84
Only null left
 
AAH3705114
View Documents Promotion Details
N/A 25 g
Each for $121.36
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10009-20-8
242.20
PZZHRSVBHRVIMI-MLHKIVSYNA-N
7009573
N[C@@H](CCCCNC(=O)C(F)(F)F)C(O)=O
C8H13F3N2O3
MFCD00037223
h-lys tfa-oh, n-6-trifluoroacetyl-l-lysine, n6-trifluoroacetyl-l-lysine, n6-trifluoroaccety-l-lysine, epsilon-tfa-lysine, nepsilon-trifluoroacetyl-l-lysine, l-lysine, n6-trifluoroacetyl, n 6-trifluoroacetyl-l-lysine, n-, a-trifluoroacetyl-l-lysine
CHEBI:61064

Specifications

∽253°C (decomposition)
C8H13F3N2O3
h-lys tfa-oh, n-6-trifluoroacetyl-l-lysine, n6-trifluoroacetyl-l-lysine, n6-trifluoroaccety-l-lysine, epsilon-tfa-lysine, nepsilon-trifluoroacetyl-l-lysine, l-lysine, n6-trifluoroacetyl, n 6-trifluoroacetyl-l-lysine, n-, a-trifluoroacetyl-l-lysine
N[C@@H](CCCCNC(=O)C(F)(F)F)C(O)=O
242.20
CHEBI:61064
+19° (c=1 in 1M HCl)
Nepsilon-Trifluoroacetyl-L-lysine
10009-20-8
MFCD00037223
PZZHRSVBHRVIMI-MLHKIVSYNA-N
(2S)-2-amino-6-[(2,2,2-trifluoroacetyl)amino]hexanoic acid
7009573
242.2
97%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only