missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Trifluoroacetyl-L-lysine, 97%

CAS: 10009-20-8 | C8H13F3N2O3 | 242.20 g/mol

Supplier:  Thermo Scientific Chemicals H3705106

Quantity 5 g

 View more versions of this product

Catalog No. AAH3705106


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10009-20-8
242.20
PZZHRSVBHRVIMI-MLHKIVSYNA-N
7009573
N[C@@H](CCCCNC(=O)C(F)(F)F)C(O)=O
C8H13F3N2O3
MFCD00037223
h-lys tfa-oh, n-6-trifluoroacetyl-l-lysine, n6-trifluoroacetyl-l-lysine, n6-trifluoroaccety-l-lysine, epsilon-tfa-lysine, nepsilon-trifluoroacetyl-l-lysine, l-lysine, n6-trifluoroacetyl, n 6-trifluoroacetyl-l-lysine, n-, a-trifluoroacetyl-l-lysine
CHEBI:61064

Specifications

∽253°C (decomposition)
10009-20-8
MFCD00037223
PZZHRSVBHRVIMI-MLHKIVSYNA-N
(2S)-2-amino-6-[(2,2,2-trifluoroacetyl)amino]hexanoic acid
242.20
CHEBI:61064
+19° (c=1 in 1M HCl)
Nepsilon-Trifluoroacetyl-L-lysine
C8H13F3N2O3
h-lys tfa-oh, n-6-trifluoroacetyl-l-lysine, n6-trifluoroacetyl-l-lysine, n6-trifluoroaccety-l-lysine, epsilon-tfa-lysine, nepsilon-trifluoroacetyl-l-lysine, l-lysine, n6-trifluoroacetyl, n 6-trifluoroacetyl-l-lysine, n-, a-trifluoroacetyl-l-lysine
N[C@@H](CCCCNC(=O)C(F)(F)F)C(O)=O
5 g
7009573
242.2
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions