accessibility menu, dialog, popup

UserName

HNOFFF(S)OHONH2

CAS RN 10009-20-8

IUPAC Name: (2S)-2-amino-6-(2,2,2-trifluoroacetamido)hexanoic acid
Synonyms: N6-(trifluoroacetyl)-L-lysine
Molecular Weight (g/mol): 242.2
Molecular Formula: C8H13F3N2O3
InChi Key: PZZHRSVBHRVIMI-MLHKIVSYNA-N
SMILES: N[C@@H](CCCCNC(=O)C(F)(F)F)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)