missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-(tert-Butoxycarbonyl)-Nalpha-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine 98.0+%, TCI America™

Catalog No. B30711G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30711G 1 g
B30715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30711G Supplier TCI America Supplier No. B30711G
Only null left

Chemical Identifiers

CAS 92122-45-7
Molecular Formula C26H32N2O6
Molecular Weight (g/mol) 468.55
MDL Number MFCD00065660
InChI Key UMRUUWFGLGNQLI-ANBDAQEENA-N
Synonym fmoc-d-lys boc-oh, nepsilon-tert-butoxycarbonyl-nalpha-9h-fluoren-9-ylmethoxy carbonyl-d-lysine, r-6-tert-butoxycarbonyl amino-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid, nepsilon-boc-nalpha-fmoc-d-lysine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid, n epsilon-tert-butoxycarbonyl-n alpha-9-fluorenylmethoxycarbonyl-d-lysine, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-t-butyl-oxycarbonyl-d-lysine, fmocdlys boc oh, 2r-6-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzfaa1330
PubChem CID 13585941
IUPAC Name (2R)-6-{[(tert-butoxy)carbonyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid
SMILES CC(C)(C)OC(=O)NCCCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 1 g
Formula Weight 468.55
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nepsilon-(tert-Butoxycarbonyl)-Nalpha-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine
TSCA No
Recommended Storage Refrigerator