missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Boc-L-lysine methyl ester hydrochloride, 98%

Catalog No. AAH6372703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6372703 1 g
AAH6372706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6372703 Supplier Thermo Scientific Chemicals Supplier No. H6372703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2389-48-2 | C12H25ClN2O4 | 296.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in methanol.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 2389-48-2
Molecular Formula C12H25ClN2O4
Molecular Weight (g/mol) 296.79
MDL Number MFCD00076959
InChI Key NANRHOPPXCBHGI-MVRPGTNWNA-N
Synonym h-lys boc-ome.hcl, h-lys boc-ome hcl, h-lys boc-ome hydrochloride, lys boc-ome.hcl, s-methyl 2-amino-6-tert-butoxycarbonyl amino hexanoate hydrochloride, methyl 2s-2-amino-6-tert-butoxycarbonyl amino hexanoate hydrochloride, pubchem18976, h-lys boc-ome?cl, lys boc-ome. hcl, h-lys boc-ome??hcl
PubChem CID 16218818
IUPAC Name methyl (2S)-2-amino-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate;hydrochloride
SMILES Cl.COC(=O)[C@@H](N)CCCCNC(=O)OC(C)(C)C

Specifications

Beilstein 5173577
Solubility Information Soluble in methanol.
Quantity 1 g
Formula Weight 296.79
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Nepsilon-Boc-L-lysine methyl ester hydrochloride
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only