missing translation for 'onlineSavingsMsg'
Learn More

Neopentyl Glycol Bis(4-aminophenyl) Ether 97.0+%, TCI America™

Catalog No. N061425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N061425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N061425G Supplier TCI America Supplier No. N061425G

Chemical Identifiers

CAS 115570-52-0
Molecular Formula C17H22N2O2
Molecular Weight (g/mol) 286.375
MDL Number MFCD00671569
InChI Key HPUJEBAZZTZOFL-UHFFFAOYSA-N
Synonym 2,2-Bis[(4-aminophenoxy)methyl]propane, 1,3-Bis(4-aminophenoxy)neopentane, 2,2-Dimethylpropane 1,3-Bis(4-aminophenyl) Ether
PubChem CID 14121182
IUPAC Name 4-[3-(4-aminophenoxy)-2,2-dimethylpropoxy]aniline
SMILES CC(C)(COC1=CC=C(C=C1)N)COC2=CC=C(C=C2)N

Specifications

Melting Point 116°C
Quantity 25 g
Formula Weight 286.38
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Neopentyl Glycol Bis(4-aminophenyl) Ether