Learn More
Invitrogen™ Neon™ NxT Electroporation System 1-Channel Tubes
Description
Neon NxT Electroporation System 1-Channel Tubes are designed specifically for use with the Neon NxT Electroporation System with 1-Channel Pipette (Cat. Nos. NEON1S, NEON1SK, NEON18S, NEON18SK). Each Neon NxT Electroporation System Kit for 25 x 2 reactions (Cat. Nos. N1025 and N10025) comes with eight 1-channel tubes, and each Neon NxT Electroporation System Kit with 1-Channel Tubes for 96 x 2 reactions (Cat. Nos. N1096 and N10096) comes with 32 1-channel tubes. This pack of additional tubes is offered for those wishing to use single tubes per reaction depending on their experimental setup or for reactions with the 1-channel pipette when predominately using the 8-channel pipette and utilizing kits that contain the 8-channel tubes.
Specifications
Specifications
| Content And Storage | Includes 24 tubes, use at room temperature under normal laboratory conditions. Refer to the Neon™ NxT Electroporation System user manual for protocol instructions. |
| Includes | Includes 24 tubes, use at room temperature under normal laboratory conditions. Refer to the Neon™ NxT Electroporation System user manual for protocol instructions. |
| Quantity | 24 Tubes |
| Cell Type | Stem Cells, Primary Cells, Hard-to-Transfect Cells |
| Format | Electroporation Pipette Tip |
| Transfection Technique | Electroporation |
| For Use With (Equipment) | Neon™ NxT Electroporation System |
| For Use With (Application) | Electroporation, Transfection |
For Research Use Only. Not for use in diagnostic procedures.