missing translation for 'onlineSavingsMsg'
Learn More

Neodymium ICP Standard, 1000 ppm Nd in 3% HNO3, Ricca Chemical

Catalog No. PND1KN100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PND1KN100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PND1KN100 Supplier Ricca Chemical Company Supplier No. PND1KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1313-97-9 , 7697-37-2 , 7732-18-5
Molecular Formula Nd2O3
Molecular Weight (g/mol) 336.48
MDL Number MFCD00011134
InChI Key PLDDOISOJJCEMH-UHFFFAOYSA-N
IUPAC Name dineodymium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Nd+3].[Nd+3]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Nd
CAS Min % 93.0, 4.62, 0.11
CAS Max % 97.0, 4.81, 0.11
Equivalence 1 mL = 1 mg Nd
Physical Form Liquid
Color Purple
pH < 2
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Neodymium ICP Standard
Show More Show Less