missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Nelfinavir mesylate

Catalog No. 376610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
376610 10 mg
376650 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 376610 Supplier Tocris Bioscience™ Supplier No. 3766/10

Potent HIV-1 protease inhibitor

Orally active human immunodeficiency virus protease inhibitor. Potently inhibits HIV-1 protease (Ki = 2 nM) in vitro. Also exhibits anti-MERS-CoV activity in Vero cells in vitro (IC50 = 0.08 μM).

Chemical Identifiers

CAS 159989-65-8
Molecular Formula C33H49N3O7S2

Specifications

Quantity 10 mg
Formula Weight 663.89
Purity >98%
Molecular Weight (g/mol) 663.889
Chemical Name or Material Nelfinavir mesylate
Recommended Storage Store at -20°C