missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Nelfinavir mesylate
$1,197.68 - $3,384.05
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC466200010
|
Thermo Scientific Chemicals
466200010 |
1 g |
N/A
|
|
|||||
|
AC466202500
|
Thermo Scientific Chemicals
466202500 |
250 mg |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 159989-65-8 | |
| C32H45N3O4S·CH4O3S |
Specifications
| 159989-65-8 | |
| Solubility aq. soln.: 4,9 g/L @25°C. Other solubilities: soluble in methanol, ethanol | |
| C32H45N3O4S·CH4O3S | |
| NQHXCOAXSHGTIA-SKXNDZRYSA-N | |
| (3S,4aS,8aS)-N-tert-butyl-2-[(2R,3R)-2-hydroxy-3-[(3-hydroxy-2-methylphenyl)formamido]-4-(phenylsulfanyl)butyl]-decahydroisoquinoline-3-carboxamide; methanesulfonic acid | |
| Nelfinavir mesylate |
| 1 g | |
| 131.0°C to 135.0°C | |
| 663.89 | |
| CS(O)(=O)=O.CC1=C(O)C=CC=C1C(=O)N[C@@H](CSC1=CC=CC=C1)[C@H](O)CN1C[C@H]2CCCC[C@H]2C[C@H]1C(=O)NC(C)(C)C | |
| 663.89 |
RUO – Research Use Only