missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nelfinavir mesylate

Catalog No. AC466200010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466200010 1 g
AC466202500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466200010 Supplier Thermo Scientific Chemicals Supplier No. 466200010

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 159989-65-8
Molecular Formula C32H45N3O4S·CH4O3S

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: 4,9 g/L @25°C. Other solubilities: soluble in methanol, ethanol
Melting Point 131.0°C to 135.0°C
Formula Weight 663.89
InChI Key NQHXCOAXSHGTIA-SKXNDZRYSA-N
SMILES CS(O)(=O)=O.CC1=C(O)C=CC=C1C(=O)N[C@@H](CSC1=CC=CC=C1)[C@H](O)CN1C[C@H]2CCCC[C@H]2C[C@H]1C(=O)NC(C)(C)C
IUPAC Name (3S,4aS,8aS)-N-tert-butyl-2-[(2R,3R)-2-hydroxy-3-[(3-hydroxy-2-methylphenyl)formamido]-4-(phenylsulfanyl)butyl]-decahydroisoquinoline-3-carboxamide; methanesulfonic acid
Molecular Weight (g/mol) 663.89
Chemical Name or Material Nelfinavir mesylate

RUO – Research Use Only