New England Biolab Ltd NEBNext Ultra™ II End Repair/dA-Tailing Module 96 reactions
Numéro de catalogue. 501047502
This module is part of the Ultra II workflow, and is optimized for use with the NEBNext Ultra II Ligation Module , for Illumina-compatible library construction. This module is also compatible with some Oxford Nanopore MinIONworkflows.
Numéro de catalogue.
501047502
Fournisseur
New England Biolab LtdCode fournisseur
E7546L
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
This module is part of the Ultra II workflow, and is optimized for use with the NEBNext Ultra II Ligation Module , for Illumina-compatible library construction. This module is also compatible with some Oxford Nanopore MinIONworkflows.