Learn More
New England Biolab Ltd NEBNext Ultra™ II RNA Library Prep Kit for Illumina 96 reactions
Numéro de catalogue. 501526375
Encompass
The Ultra II RNA Library Prep Kit for Illumina is designed for non-directional (non-strand-specific) RNA library construction, and delivers significantly increased sensitivity and specificity from your RNA-seq experiments, from ever-decreasing amounts of input RNA. In conjunction with ribosomal RNA (rRNA) depletion or poly(A) mRNA enrichment, the kit enables the production of high quality libraries from 5 ng or 10 ng of Total RNA, respectively, up to 1g.
Numéro de catalogue. 501526375 Fournisseur New England Biolab Ltd Code fournisseur E7770L
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
The Ultra II RNA Library Prep Kit for Illumina is designed for non-directional (non-strand-specific) RNA library construction, and delivers significantly increased sensitivity and specificity from your RNA-seq experiments, from ever-decreasing amounts of input RNA. In conjunction with ribosomal RNA (rRNA) depletion or poly(A) mRNA enrichment, the kit enables the production of high quality libraries from 5 ng or 10 ng of Total RNA, respectively, up to 1g.