New England Biolab Ltd NEBNext Single Cell/Low Input cDNA Synthesis and Amplification Module 24 reactions
Numéro de catalogue. NC1517720
The NEBNext Single Cell/Low Input cDNA Synthesis and Amplification Module uses a template switching method to generate full length cDNAs directly from single cells or 2 pg - 200 ng RNA. This unique workflow enables generation of high quality sequencing data from a broad range of inputs, and superior transcript detection, while providing reliably consistent performance
Numéro de catalogue.
NC1517720
Fournisseur
New England Biolab LtdCode fournisseur
E6421S
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
The NEBNext Single Cell/Low Input cDNA Synthesis and Amplification Module uses a template switching method to generate full length cDNAs directly from single cells or 2 pg - 200 ng RNA. This unique workflow enables generation of high quality sequencing data from a broad range of inputs, and superior transcript detection, while providing reliably consistent performance