Learn More
New England Biolab Ltd NEBExpress Cell-Free E. coli Protein Synthesis System 10 reactions
Numéro de catalogue. 502024007
Encompass
The NEBExpress Cell-free E. coli Protein Synthesis System is an extract-based transcription/translation system derived from E. coli cells that have been engineered for high in vitro synthesis performance. It is designed to synthesize proteins ranging in size from 17-230 kDa in just 2-4 hours under the control of T7 RNA Polymerase.
Numéro de catalogue. 502024007 Fournisseur New England Biolab Ltd Code fournisseur E5360S
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
The NEBExpress Cell-free E. coli Protein Synthesis System is an extract-based transcription/translation system derived from E. coli cells that have been engineered for high in vitro synthesis performance. It is designed to synthesize proteins ranging in size from 17-230 kDa in just 2-4 hours under the control of T7 RNA Polymerase.