New Era Pump Systems Inc NE-9000B Programmable Peristaltic Pump
Numéro de catalogue. NC1416046
This pump is used for higher flow rates & larger dispense volumes. NE-9000 with 2 roller pumping head & 3/16" ID tubing Flow rate ranges: 0.035 mL/min to 775.2 mL/min. Includes PERI-HEAD-KIT-YB2.
Numéro de catalogue.
NC1416046
Fournisseur
New Era Pump Systems IncCode fournisseur
NE9000B
This pump is used for higher flow rates & larger dispense volumes. NE-9000 with 2 roller pumping head & 3/16" ID tubing Flow rate ranges: 0.035 mL/min to 775.2 mL/min. Includes PERI-HEAD-KIT-YB2.